siRNA Details
| Gene Symbol: POU3F3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006236.2 | NP_006227.1 | 1 | 4482 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR599621.1 | - | 1 | 968 | Chr. 2 - 104857582 - 104858574 [+] on Build GRCh38 | |

POU3F3
Genomic Map