siRNA Details
| Gene Symbol: CDKN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF488411.1 | - | 831 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDKN2B| Gene Symbol: CDKN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF488411.1 | - | 831 | |||