siRNA Details
| Gene Symbol: PKD1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017011798.1 | XP_016867287.1 | 1 | 2892 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK001279.1 | - | 1 | 1582 | Chr. 7 - 47955352 - 47957314 [-] on Build GRCh38 | |

PKD1L1
Genomic Map