siRNA Details
| Gene Symbol: PKD1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017011798.1 | XP_016867287.1 | 1 | 2553 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PKD1L1| Gene Symbol: PKD1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017011798.1 | XP_016867287.1 | 1 | 2553 | ||