siRNA Details
| Gene Symbol: LOC105379305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CR936668.1 | - | 1 | 636 | ||
| Gene Symbol: LOC101929355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017020910.1 | XP_016876399.1 | 1 | 470 | ||

LOC105379305
Genomic Map