siRNA Details
| Gene Symbol: CD200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005247481.4 | XP_005247538.1 | 6 | 3247 | ||
| XM_005247485.2 | XP_005247542.1 | 5 | 3188 | |||
| XM_005247486.2 | XP_005247543.1 | 4 | 2685 | |||
| XM_005247488.2 | XP_005247545.1 | 4 | 2790 | |||
| XM_017006452.1 | XP_016861941.1 | 6 | 3574 | |||
| XM_017006453.1 | XP_016861942.1 | 4 | 2840 | |||
| XM_017006457.1 | XP_016861946.1 | 5 | 2872 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK001097.1 | - | 2 | 2089 | Chr. 10 - 29653015 - 29675289 [-] on Build GRCh38 | |

CD200
Genomic Map