siRNA Details
| Gene Symbol: NDE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017023349.1 | XP_016878838.1 | 9 | 2648 | ||
| XM_017023351.1 | XP_016878840.1 | 10 | 2727 | |||
| XM_017023353.1 | XP_016878842.1 | 9 | 2780 | |||
| XM_017023354.1 | XP_016878843.1 | 8 | 2496 | |||
| XM_017023356.1 | XP_016878845.1 | 9 | 2684 | |||
| GenBank mRNA | AK123247.1 | - | 2 | 1742 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA941555.1 | - | 1 | 48 | Chr. 16 - 15701280 - 15701837 [+] on Build GRCh38 | |

NDE1
Genomic Map