siRNA Details
| Gene Symbol: HOXB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011524707.2 | XP_011523009.1 | 1 | 121 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA746255.1 | - | 1 | 122 | Chr. 17 - 48574311 - 48574798 [-] on Build GRCh38 | |

HOXB3
Genomic Map