siRNA Details
| Gene Symbol: UBE2D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015983.3 | NP_057067.1 | 7 | 3264 | ||
| XM_006715734.3 | XP_006715797.1 | 8 | 2851 | |||
| XM_011515420.2 | XP_011513722.1 | 8 | 3357 | |||
| XM_011515422.2 | XP_011513724.1 | 6 | 3201 | |||
| XM_011515423.2 | XP_011513725.1 | 5 | 2635 | |||
| XM_017012292.1 | XP_016867781.1 | 9 | 2944 | |||
| XM_017012295.1 | XP_016867784.1 | 6 | 2698 | |||
| GenBank mRNA | AK056566.1 | - | 1 | 1881 | ||

UBE2D4
Genomic Map