siRNA Details
| Gene Symbol: DISC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL049973.2 | - | 1 | 1387 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DISC1| Gene Symbol: DISC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL049973.2 | - | 1 | 1387 | ||