siRNA Details
| Gene Symbol: SSH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006722149.3 | XP_006722212.1 | 1 | 2658 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK127747.1 | - | 1 | 733 | Chr. 13 - 47932299 - 47935053 [-] on Build GRCh38 | |

SSH2
Genomic Map