siRNA Details
Gene Symbol: LOC101929680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110260.1 | 5 | 676 | |||
GenBank mRNA | BC029452.1 | - | 371 | |||
KM096550.1 | - | 4 | 492 | |||
KM096551.1 | - | 4 | 492 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110260.1 | 5 | 676 | |||
GenBank mRNA | BC029452.1 | - | 371 | |||
KM096550.1 | - | 4 | 492 | |||
KM096551.1 | - | 4 | 492 |