siRNA Details
| Gene Symbol: LOC101929680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC029452.1 | - | 307 | |||
| KM096550.1 | - | 3, 4 | 428 | |||
| KM096551.1 | - | 3, 4 | 428 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929680| Gene Symbol: LOC101929680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC029452.1 | - | 307 | |||
| KM096550.1 | - | 3, 4 | 428 | |||
| KM096551.1 | - | 3, 4 | 428 | |||