siRNA Details
| Gene Symbol: SEMA6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK096337.1 | - | 1 | 504 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AB002438.1 | - | 1 | 1374 | Chr. 5 - 116468957 - 116470410 [-] on Build GRCh38 | |

SEMA6A
Genomic Map