siRNA Details
| Gene Symbol: LOC440934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136638.1 | 2 | 1126 | |||
| NR_136639.1 | 2 | 830 | ||||
| NR_136640.1 | 2 | 395 | ||||
| NR_136641.1 | 2 | 574 | ||||
| GenBank mRNA | BC008048.2 | - | 2 | 1125 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC440934| Gene Symbol: LOC440934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136638.1 | 2 | 1126 | |||
| NR_136639.1 | 2 | 830 | ||||
| NR_136640.1 | 2 | 395 | ||||
| NR_136641.1 | 2 | 574 | ||||
| GenBank mRNA | BC008048.2 | - | 2 | 1125 | ||