siRNA Details
| Gene Symbol: LOC440934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136638.1 | 1 | 825 | |||
| NR_136642.1 | 1 | 825 | ||||
| GenBank mRNA | BC008048.2 | - | 1 | 824 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC440934| Gene Symbol: LOC440934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136638.1 | 1 | 825 | |||
| NR_136642.1 | 1 | 825 | ||||
| GenBank mRNA | BC008048.2 | - | 1 | 824 | ||