siRNA Details
| Gene Symbol: LRRC8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC053565.1 | - | 1 | 487 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ717287.1 | - | 1 | 486 | Chr. 1 - 89584150 - 89584917 [-] on Build GRCh38 | |

LRRC8B
Genomic Map