siRNA Details
| Gene Symbol: RFK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK123933.1 | - | 1 | 1756 | ||
| AK307079.1 | - | 2 | 859 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RFK| Gene Symbol: RFK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK123933.1 | - | 1 | 1756 | ||
| AK307079.1 | - | 2 | 859 | |||