siRNA Details
| Gene Symbol: LCMT1-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK002209.1 | - | 1 | 93 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AU140974.1 | - | 1 | 93 | Chr. 16 - 25145492 - 25146248 [-] on Build GRCh38 | |

LCMT1-AS2
Genomic Map