siRNA Details
Gene Symbol: KRTAP4-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030976.1 | NP_112238.1 | 1 | 769 | ||
GenBank mRNA | AJ406938.1 | - | 1 | 468 | ||
AJ406945.1 | - | 1 | 733 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KRTAP4-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030976.1 | NP_112238.1 | 1 | 769 | ||
GenBank mRNA | AJ406938.1 | - | 1 | 468 | ||
AJ406945.1 | - | 1 | 733 |