siRNA Details
| Gene Symbol: KRTAP4-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030976.1 | NP_112238.1 | 1 | 627 | ||
| GenBank mRNA | AJ406938.1 | - | 1 | 326 | ||
| AJ406945.1 | - | 1 | 591 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP4-6| Gene Symbol: KRTAP4-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030976.1 | NP_112238.1 | 1 | 627 | ||
| GenBank mRNA | AJ406938.1 | - | 1 | 326 | ||
| AJ406945.1 | - | 1 | 591 | |||