siRNA Details
| Gene Symbol: LINC00564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047494.1 | 1 | 517 | |||
| GenBank mRNA | BC039360.1 | - | 1 | 517 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00564| Gene Symbol: LINC00564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047494.1 | 1 | 517 | |||
| GenBank mRNA | BC039360.1 | - | 1 | 517 | ||