siRNA Details
| Gene Symbol: PPP1R2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC026109.1 | - | 1 | 417 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI459936.1 | - | 1 | 417 | Chr. 21 - 35887067 - 35887684 [+] on Build GRCh38 | |

PPP1R2P2
Genomic Map