siRNA Details
| Gene Symbol: PRKG1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC039504.2 | - | 4 | 749 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM479024.1 | - | 4 | 761 | Chr. 10 - 52252147 - 52297917 [-] on Build GRCh38 | |

PRKG1-AS1
Genomic Map