siRNA Details
| Gene Symbol: LOC101927606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131977.1 | 2 | 669 | |||
| GenBank mRNA | BC039507.1 | - | 2 | 668 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000567168.1 | - | 2 | 669 | Chr. 18 - 74409448 - 74413209 [+] on Build GRCh38 | |

LOC101927606
Genomic Map