siRNA Details
| Gene Symbol: SEPT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011553.3 | NP_001011553.2 | 13 | 3569 | ||
| NM_001242956.1 | NP_001229885.1 | 12 | 3464 | |||
| NM_001788.5 | NP_001779.3 | 14 | 3572 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEPT7| Gene Symbol: SEPT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011553.3 | NP_001011553.2 | 13 | 3569 | ||
| NM_001242956.1 | NP_001229885.1 | 12 | 3464 | |||
| NM_001788.5 | NP_001779.3 | 14 | 3572 | |||