siRNA Details
| Gene Symbol: LINC01109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK124684.1 | - | 1 | 888 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB540288.1 | - | 1 | 339 | Chr. 8 - 76404607 - 76405091 [+] on Build GRCh38 | |

LINC01109
Genomic Map