siRNA Details
| Gene Symbol: LOC101927053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110209.1 | 3 | 2156 | |||
| GenBank mRNA | BC068584.1 | - | 3 | 2156 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927053| Gene Symbol: LOC101927053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110209.1 | 3 | 2156 | |||
| GenBank mRNA | BC068584.1 | - | 3 | 2156 | ||