siRNA Details
| Gene Symbol: ZSWIM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AW997845.1 | - | 1 | 180 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC082763.1 | - | 22 | 4534 | Chr. 10 - 73785656 - 73801793 [-] on Build GRCh38 | |

ZSWIM8
Genomic Map