siRNA Details
| Gene Symbol: ZNF391 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076781.2 | NP_001070249.1 | 3 | 2929 | ||
| NM_001322288.1 | NP_001309217.1 | 4 | 3153 | |||
| NM_001322289.1 | NP_001309218.1 | 4 | 3008 | |||
| NM_001322293.1 | NP_001309222.1 | 3 | 2897 | |||
| XM_005249082.3 | XP_005249139.1 | 3 | 2714 | |||
| XM_006715086.2 | XP_006715149.1 | 3 | 2655 | |||
| XM_011514569.1 | XP_011512871.1 | 3 | 2637 | |||
| GenBank mRNA | BC035154.2 | - | 1 | 872 | ||
| BM906507.1 | - | 1 | 591 | |||

ZNF391
Genomic Map