siRNA Details
| Gene Symbol: OR11A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ459854.1 | - | 1 | 61 | ||
| Gene Symbol: OR5V1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011514929.2 | XP_011513231.1 | 1 | 57 | ||
| XM_011514930.2 | XP_011513232.1 | 1 | 29 | |||

OR11A1
Genomic Map