siRNA Details
| Gene Symbol: LOC101928201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110390.1 | 3 | 315 | |||
| GenBank mRNA | BC038195.1 | - | 3 | 315 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928201| Gene Symbol: LOC101928201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110390.1 | 3 | 315 | |||
| GenBank mRNA | BC038195.1 | - | 3 | 315 | ||