siRNA Details
| Gene Symbol: LOC100507388 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC047744.1 | - | 1 | 256 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ956614.1 | - | 1 | 255 | Chr. 4 - 74965281 - 74970123 [-] on Build GRCh38 | |

LOC100507388
Genomic Map