siRNA Details
| Gene Symbol: LOC100507388 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC047744.1 | - | 3 | 897 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507388| Gene Symbol: LOC100507388 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC047744.1 | - | 3 | 897 | ||