siRNA Details
| Gene Symbol: LOC100507388 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121617.1 | 2 | 150 | |||
| GenBank mRNA | AK027257.1 | - | 2 | 150 | ||
| BC047744.1 | - | 1 | 557 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ956614.1 | - | 1 | 556 | Chr. 4 - 74965281 - 74970123 [-] on Build GRCh38 | |

LOC100507388
Genomic Map