siRNA Details
| Gene Symbol: LOC101927531 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC040862.2 | - | 2 | 588 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM800740.1 | - | 2 | 600 | Chr. 12 - 126359524 - 126360884 [+] on Build GRCh38 | |

LOC101927531
Genomic Map