siRNA Details
| Gene Symbol: CDKL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA742987.1 | - | 1 | 170 | ||
| DA508790.1 | - | 1 | 124 | |||
| HY355743.1 | - | 1 | 163 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDKL1| Gene Symbol: CDKL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA742987.1 | - | 1 | 170 | ||
| DA508790.1 | - | 1 | 124 | |||
| HY355743.1 | - | 1 | 163 | |||