siRNA Details
Gene Symbol: POU3F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006236.2 | NP_006227.1 | 1 | 551 | ||
GenBank mRNA | CX873380.1 | - | 1 | 491 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: POU3F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006236.2 | NP_006227.1 | 1 | 551 | ||
GenBank mRNA | CX873380.1 | - | 1 | 491 |