siRNA Details
| Gene Symbol: BLACAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103783.1 | 2 | 260 | |||
| GenBank mRNA | AK022594.1 | - | 1 | 12 | ||
| BX640973.1 | - | 1 | 639 | |||
| DA703157.1 | - | 2 | 260 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BLACAT1| Gene Symbol: BLACAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103783.1 | 2 | 260 | |||
| GenBank mRNA | AK022594.1 | - | 1 | 12 | ||
| BX640973.1 | - | 1 | 639 | |||
| DA703157.1 | - | 2 | 260 | |||