siRNA Details
Gene Symbol: LOC146795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135222.1 | 1, 2 | 1330 | |||
GenBank mRNA | AK057377.1 | - | 1, 2 | 1202 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC146795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135222.1 | 1, 2 | 1330 | |||
GenBank mRNA | AK057377.1 | - | 1, 2 | 1202 |