siRNA Details
| Gene Symbol: LOC100130027 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK127225.1 | - | 1 | 1353 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100130027| Gene Symbol: LOC100130027 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK127225.1 | - | 1 | 1353 | ||