siRNA Details
| Gene Symbol: WIPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005257083.2 | XP_005257140.1 | 8 | 7182 | ||
| XM_005257084.2 | XP_005257141.1 | 8 | 7140 | |||
| XM_011524412.1 | XP_011522714.1 | 7 | 7081 | |||
| XM_011524413.1 | XP_011522715.1 | 7 | 7062 | |||
| GenBank mRNA | BC030561.1 | - | 1 | 1175 | ||

WIPF2
Genomic Map