siRNA Details
| Gene Symbol: SLC19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017028444.1 | XP_016883933.1 | 7 | 4304 | ||
| XM_017028446.1 | XP_016883935.1 | 6 | 4164 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC19A1| Gene Symbol: SLC19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017028444.1 | XP_016883933.1 | 7 | 4304 | ||
| XM_017028446.1 | XP_016883935.1 | 6 | 4164 | |||