siRNA Details
| Gene Symbol: LOC100507654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC034289.1 | - | 1, 2 | 73 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG721018.1 | - | 1, 2 | 78 | Chr. 13 - 100537323 - 100580113 [-] on Build GRCh38 | |

LOC100507654
Genomic Map