siRNA Details
| Gene Symbol: LOC100507654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC034289.1 | - | 4 | 2055 | ||
| Gene Symbol: GGACT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011521129.2 | XP_011519431.1 | 1 | 1850 | ||

LOC100507654
Genomic Map