siRNA Details
| Gene Symbol: LOC115110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037844.2 | 6, 7 | 4262 | |||
| GenBank mRNA | AK021767.1 | - | 2, 3 | 1272 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC115110| Gene Symbol: LOC115110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037844.2 | 6, 7 | 4262 | |||
| GenBank mRNA | AK021767.1 | - | 2, 3 | 1272 | ||