siRNA Details
| Gene Symbol: LOC115110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037844.2 | 6 | 4226 | |||
| GenBank mRNA | AK021767.1 | - | 2 | 1236 | ||
| AL359943.1 | - | 2 | 3277 | |||
| CR749717.1 | - | 2 | 3277 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA290908.1 | - | 3 | 474 | Chr. 1 - 2550572 - 2553175 [-] on Build GRCh38 | |

LOC115110
Genomic Map