siRNA Details
| Gene Symbol: LINC01016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC034285.1 | - | 1 | 231 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01016| Gene Symbol: LINC01016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC034285.1 | - | 1 | 231 | ||