siRNA Details
| Gene Symbol: LINC01016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038989.1 | 3 | 677 | |||
| GenBank mRNA | AK057709.1 | - | 3 | 675 | ||
| BC034285.1 | - | 1 | 737 | |||
| BC042121.1 | - | 2 | 510 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01016| Gene Symbol: LINC01016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038989.1 | 3 | 677 | |||
| GenBank mRNA | AK057709.1 | - | 3 | 675 | ||
| BC034285.1 | - | 1 | 737 | |||
| BC042121.1 | - | 2 | 510 | |||