siRNA Details
| Gene Symbol: LOC101927870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110141.1 | 4 | 2166 | |||
| GenBank mRNA | BC041378.2 | - | 4 | 2166 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927870| Gene Symbol: LOC101927870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110141.1 | 4 | 2166 | |||
| GenBank mRNA | BC041378.2 | - | 4 | 2166 | ||